Anti-Antiplasmin (serine (or Cysteine) peptidase inhibitor clade F member 2) Rabbit Polyclonal Antibody
Catalog # 89295-748
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Antiplasmin (serine (or Cysteine) peptidase inhibitor clade F member 2)
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Size:500 μg
- Gene ID:18816
- Storage buffer:PBS, pH 7.4; 1% BSA, 0.01% Thimerosal
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:2mg/ml
- Purification:Affinity purified by Protein A.
- Cat. no.:89295-748
- Supplier No.:GTX79826
Specifications
About this item
Rabbit Polyclonal antibody to antiplasmin (HRP)
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse