Anti-PAI1 Mouse Monoclonal Antibody [clone: H4B3]
Catalog # 89333-574
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Plasminogen Activator Inhibitor 1
- Antigen symbol:PAI1
- Clonality:Monoclonal
- Clone:H4B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Size:100 μg
- Neutralisation:Yes
- Gene ID:18787
- Antigen synonyms:serpin family E member 1|PLANH1|PAI1|PAI|SERPINE1|PAI-1
- Storage buffer:0.05M Sodium Phosphate; 0.15M NaCl; pH 7.4
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:2mg/ml
- Purification:Affinity purified by Protein A.
- Cat. no.:89333-574
- Supplier No.:GTX79706
Specifications
About this item
Inhibitory Mouse Monoclonal antibody to mouse PAI-1 [H4B3]
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: PAI1
Clonality: Monoclonal
Clone: H4B3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Pig, Rat