Anti-MGC39633 Rabbit Polyclonal Antibody
Catalog # 89288-570
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Coiled-coil Domain containing 112
- Antigen symbol:MGC39633
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog
- Western blot:Yes
- Size:100 μg
- Gene ID:153733
- Antigen synonyms:MBC1
- Storage buffer:Phosphate buffered saline (PBS)
- Storage temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Protein A Purified Total IgG
- Cat. no.:89288-570
- Supplier No.:GTX46594
Specifications
About this item
Rabbit polyclonal antibody to MGC39633 (N-terminal)
Recommended Dilutions: For Western Blot: Use at 1.25 ug/ml. For IHC-P: Use at 4-8 ug/mL.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: MGC39633
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog