Anti-TMPRSS11D Rabbit Polyclonal Antibody
Catalog # 89322-992
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane Protease Serine 11D
- Antigen symbol:TMPRSS11D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:9407
- Antigen synonyms:Za11|IL-21|HAT|CVID11
- Storage buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.62 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human TMPRSS11D. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89322-992
- Supplier No.:GTX117370
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TMPRSS11D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human