Anti-RIPK1 Rabbit Polyclonal Antibody
Catalog # 89320-672
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Receptor (Tnfrsf)-Interacting Serine-Threonine Kinase 1
- Antigen symbol:RIPK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:8737
- Antigen synonyms:RIP1|RIP
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human RIP. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89320-672
- Supplier No.:GTX111235
Specifications
About this item
Rabbit Polyclonal antibody to RIP (receptor (TNFRSF)-interacting serine-threonine kinase 1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: RIPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human