Anti-AKT2 Rabbit Polyclonal Antibody
Catalog # 89320-000
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:V-Akt Murine Thymoma Viral Oncogene Homolog 2
- Antigen symbol:AKT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:25778
- Antigen synonyms:RAC-BETA|PKBB|PRKBB|PKBBETA|HIHGHH
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human RIPK5. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89320-000
- Supplier No.:GTX107925
Specifications
About this item
Rabbit polyclonal antibody to RIPK5 (receptor interacting protein kinase 5)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat