Order Entry
Canada
Orders LinkContactUsLinkComponent
Anti-MAPK1 Rabbit Polyclonal Antibody

Anti-MAPK1 Rabbit Polyclonal Antibody

100 μl
Catalog # 89319-058
Supplier:   Genetex
Supplier #:  GTX103792
Anti-MAPK1 Rabbit Polyclonal Antibody
Catalog # 89319-058
Supplier:  Genetex
Anti-MAPK1 Rabbit Polyclonal Antibody
Anti-MAPK1 Rabbit Polyclonal Antibody

100 μl

Price
$949.51
Log in for availability
Each (100µl)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Rabbit Polyclonal antibody to Creatine kinase (muscle) (creatine kinase, muscle)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat

Specifications
  • Size:
    100 μl
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Purified by antigen-affinity chromatography.
  • Antibody type:
    Primary
  • Isotype:
    IgG
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Host:
    Rabbit
  • Concentration:
    0.71mg/ml
  • Antigen name:
    Mitogen-Activated Protein Kinase 1
  • Cat. No.:
    89319-058
  • Reactivity:
    Monkey|Human|Dog|Rat
  • Gene ID:
    1158
  • Antigen synonyms:
    P42MAPK|PRKM2|ERK2|PRKM1|MAPK2|ERK|ERT1|p38|p41mapk|ERK-2|p40|p42-MAPK|p41
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human Creatine kinase (muscle). The exact sequence is proprietary.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    MAPK1
  • Storage buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.