Anti-QPRT Rabbit Polyclonal Antibody
Catalog # 89318-730
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Quinolinate Phosphoribosyltransferase
- Antigen symbol:QPRT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:9263
- Antigen synonyms:QPRT|QPRTase|Quinolinate phosphoribosyltransferase.|Nicotinate nucleotide pyrophosphorylase carboxylating|QAPRTase|Quinolinate phosphoribosyltransferase [decarboxylating]|Nicotinate-nucleotide pyrophosphorylase [carboxylating]|NADC_HUMAN
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.91mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human DRAK1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89318-730
- Supplier No.:GTX102394
Specifications
About this item
Rabbit Polyclonal antibody to DRAK1 (serine/threonine kinase 17a)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: QPRT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human