Anti-RAB17 Rabbit Polyclonal Antibody
Catalog # 89318-728
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-17
- Antigen symbol:RAB17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:9263
- Antigen synonyms:BOFS|AP2TF|AP-2|AP-2alpha|TFAP2
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.84mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the C-terminus region of human DRAK1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89318-728
- Supplier No.:GTX102375
Specifications
About this item
Rabbit polyclonal antibody to DRAK1 (serine/threonine kinase 17a)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: RAB17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human