Neulog Environmental Bundle
Product Details & Documents
A set of six NeuLog sensors that are great for use in environmental science classrooms.
- Collect data in the classroom or remotely
- Compatible with PC, Mac, and chromebook
- Free software available for download
- Connect multiple sensors simultaneously to a single battery or connectivity module
The NeuLog probeware system is a complete data collection platform perfect for classroom use. Data are sent from the sensors to any computer via USB, bluetooth, or WiFi. A rechargeable battery makes remote data collection easy: the sensors will store the data for pre-defined experiments until they are uploaded to a computer later. The free software is powerful and easy to use.
This bundle of sensors supports experiments in air, water, and soil quality.
Ordering information: Required but not included: NeuLog USB module or NeuLog bluetooth module (which also provide power for the sensors), or a NeuLog WiFi module plus a NeuLog rechargeable battery.
Delivery information: Bundle includes one each of the following sensors: Temperature, oxygen, pH, conductivity, soil moisture, and turbidity.
Caution: For educational use only.
Specifications
- Description:NeuLog environmental bundle