Two-Stroke Diesel Engine Model
Catalog # 470157-714
Supplier: Wards
Specifications
- Description:Two-Stroke Diesel Engine Model
- Cat. no.:470157-714
- Supplier No.:PH0490
Specifications
About this item
This model shows the internal structure and operating principles of an air cooled two-stroke engine. All parts are aluminum and plastic. Ignition is shown via a small bulb. Carburetor and fuel supply are also sectioned. Hand crank is provided for manual operation. Mounted on a base.