Orders LinkContactUsLinkComponent
Light Box and Optical Set
Light Box and Optical Set
Light box and optical set
Catalog # 470157-646
Supplier:   Wards
Supplier #:  PH0615
Light Box and Optical Set
Catalog # 470157-646
Supplier:  Wards
Light Box and Optical Set
Light Box and Optical Set
Light box and optical set
Price
$432.30
Log in for availability
Each
Product Details & Documents
This kit is very useful for reflection, refraction, and color mixing experiments.
  • Supplied with working instructions
  • Power supply not included
The light box consists of a light source 12 V, 24 W lamp, producing convergent, divergent or parallel beam. On one end, an adjustable cylindrical convex lens (for parallel beam) and on the other end, a triple aperture system for color mixing experiments (lateral aperture are provided with adjustable hinged mirror). All apertures in the box are provided with vertical channels to hold slit plates and color filters. The optical set includes acrylic blocks, set of three mirrors, 2 slit black plates, one with three narrow slits at one end and one narrow slit at the other end. Another black plate with four narrow slits at one end, one wide slit at the other end and set of seven mounted color filters.
Ordering information: 12 VDC power supply is required but not included.
Specifications
  • Description:
    Light box and optical set