ContactUsLinkComponent
Classification of Chemicals: Elements, Compounds and Mixtures
Classification of Chemicals: Elements, Compounds and Mixtures
Catalog # 470165-506
Supplier:  Ward's Science
undefined
Classification of Chemicals: Elements, Compounds and Mixtures
Catalog # 470165-506
Supplier:  Ward's Science
Supplier Number:  IS2513
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Classification of Chemicals: Elements, Compounds and Mixtures
  • No. of Lab Groups:
    15
  • Cat. no.:
    470165-506
  • Includes:
    Teacher ‘s Guide and Student Copymasters
  • Supplier No.:
    IS2513
Specifications
About this item
In the Classification of Chemicals: Elements, Compounds and Mixtures kit, students perform three labs to learn about chemical classification.
  • Students Learn Chemical Classification through Three Labs
  • Kit Covers Elements, Compounds, and Mixtures
  • Materials Suppled
The three major classes of matter are elements, compounds, and mixtures.
The lab activities to help understand the differences between the three.
Ordering information: This kit includes toluidine blue, crystal violet, safranin, chromatographic dye mixture, ethanol, copper chloride, iron metal strips, chromatography paper sheets, and capillary tubes.