Oil for Belt Driven Pumps, HyVac
Supplier: Avantor
Certificates
Certificates
About this item
Ideal for belt driven vacuum pumps
- Low vapor pressure
- Maintains oil seal
- Lubricates pump thoroughly
- Heat transfer efficiently
They resists absorption of condensates and are also thermally stable.
Frequently Bought Together
American White Cross First Aid® Assorted Designer Bandages, DUKAL™ Corporation
Supplier: Dukal
$12.62 to $151.06
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$135.75 to $468.77
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$277.09 to $740.95
Sol-Vet® Standard Insulin Syringes (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM MEDICAL
$27.21 to $266.64