Order Entry
Canada
ContactUsLinkComponent
Nitrocellulose Membrane, 0.45 µm, Bio-Rad
Nitrocellulose Membrane, 0.45 µm, Bio-Rad
Catalog # 101643-796
Supplier:  Bio-Rad
Nitrocellulose Membrane, 0.45 µm, Bio-Rad
Catalog # 101643-796
Supplier:  Bio-Rad
Supplier Number:  1620145

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Nitrocellulose membranes, sheets, pore size 0.45 µm
  • Dimensions:
    7 x 8.4 cm
  • Size:
    Pack of 10
  • Compatible with:
    Mini Trans-Blot®; Trans-Blot® SD/Mini-PROTEAN® gels
  • Cat. no.:
    101643-796
  • Binding Capacity:
    80–100 μg/cm²
  • Supplier No.:
    1620145

Specifications

About this item

Bio-Rad 0.45 µm pure nitrocellulose membrane is recommended for analytical blotting, including western blotting, dot/slot blot screening, northern blotting, and Southern blotting

Precut membrane/blot absorbent filter paper sandwiches, precut membrane sheets and membrane rolls are available in a range of sizes.

0.45 µm nitrocellulose has a high binding capacity for proteins and nucleic acids >500 Da. For short target polypeptides and nucleic acids <500 Da, Bio-Rad 0.2 µm nitrocellulose is recommended.