Order Entry
Canada
ContactUsLinkComponent
 
Gene Pulser® Electroporation Buffer, Bio-Rad
  100447-022
 :  Bio-Rad
 :  
Gene Pulser® Electroporation Buffer, Bio-Rad
  100447-022
 :  Bio-Rad
 :  1652677
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Gene Pulser® Electroporation Buffer
  • Shelf life:
    6 months from date of purchase
  • Storage:
    4°C
  • Cat. no.:
    100447-022
  • Volume:
    30 ml
  • Supplier No.:
    1652677

 

 

Gene Pulser® Electroporation Buffer is formulated to minimize cell mortality while ensuring efficient delivery of nucleic acids

  • Works with most electroporators, including Bio-Rad's Gene Pulser Xcell™ Electroporation Systems and MicroPulser™ Electroporator
  • Delivers siRNA, DNA, and other biomolecules
  • Improves efficiency of transfection and maintains cell viability after electroporation
  • Can be used for transfection in single cuvettes or in multiwell electroporation plates

This universal buffer is designed for electroporating mammalian cells, including primary cells and difficult-to-transfect cells.