Human Recombinant CNTF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3.1×10³ units/mg
- Gene ID:P26441
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:CNTF
- Purity:≥95%
- Molecular weight:22.9 kDa
- Sequence:MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together