Human Recombinant TNFR-1 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Tumor necrosis factor receptor type 1 (TNFR-1) is expressed in most tissues and is activated by soluble and membrane-bound tumor necrosis factor alpha (TNF-α). TNFR-1 activates NF-κB and MAPK pathways to induce inflammation, promote apoptotic cell death, inhibit tumorigenesis, and inhibit viral replication.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity Assay: Neutralization of human TNF alpha induced L929 cell cytolysis, ED₅₀ ≤100 ng/ml, Bioactivity ≥1.0×10⁴ units/mg
- Gene ID:P19438
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:TNFRSF1A|TNFR55|CD120a|TNFAR
- Protein/peptide name:TNFR-1
- Purity:≥95%
- Molecular weight:18.3 kDa
- Sequence:MDSVCPQGKY IHPQNNSICC TKCHKGTYLY NDCPGPGQDT DCRECESGSF TASENHLRHC LSCSKCRKEM GQVEISSCTV DRDTVCGCRKN QYRHYWSENL FQCFNCSLCL NGTVHLSCQE KQNTVCTCHA GFFLRENECV SCSNCKKSLE CTKLCLPQI EN
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together