Human Recombinant EGF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR). EGF is overexpressed in multiple tumor cell lines and promotes resistance to chemotherapy and radiation treatments.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mg
- Gene ID:P01133
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Urogastrone|URG
- Protein/peptide name:EGF
- Purity:≥95%
- Molecular weight:6.2 kDa
- Sequence:NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Related Information
Certificates
Search by Lot NumberLiterature reference - PubMed ID 21368213 Literature reference - PubMed ID 23070115 Literature reference - PubMed ID 24276241 Literature reference - PubMed ID 27030425 Literature reference - PubMed ID 27634936
Additional Product Details
More
Shenandoah_Biotechnology_Product_Catalog
Product Literature
Frequently Bought Together