Human Recombinant BAFF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
B cell-activating factor (BAFF), or B lymphocyte stimulator (BLyS), is a type II member of the tumor necrosis factor (TNF) superfamily. BAFF is expressed as a transmembrane protein on T cells, macrophages, and dendritic cells. The transmembrane domain of BAFF can also be cleaved to produce a soluble protein fragment.
- High quality
- Low endotoxin
- Affordable
BAFF binds to the TNF receptors known as B cell maturation antigen (BCMA), transmembrane activator and CAML interactor (TACI), and BAFF receptor (BAFFR). BAFF is important for the survival and maturation of peripheral B cells. Human BAFF shows activity on mouse splenocytes.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q9Y275
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:TALL|THANK|BLyS
- UniProtKB:Q9Y275
- Protein/peptide name:BAFF
- Purity:≥90%
- Molecular weight:18.5 kDa
- Sequence:MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together