Komet, Magnetic Stirring Bars, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Generate higher torque than conventional stirring bars with Komet Stir Bars.
- Magnetic power two to three times higher than that of conventional bars
- Magnetic properties remain for many years
Komet magnetic stirring bars made from SmCo (Samarium Cobalt) have a PTFE coating which means they can be sterilised. Particularly recommended for high speed stirring, liquids with a high viscosity and stirring in vessels where there is a large distance between stirrer and stirring bar, such as measuring cyliners.
Customers Who Bought This Also Bought