Specifications
- Antibody type:Primary
- Antigen symbol:DJ-1/PARK7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Size:500 µg
- Epitope:N-Terminal domain
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:PARK7
- Immunogen:A synthetic peptide (ASKRALVILAKGAEE-C) corresponding to human DJ-1 [2-16] in the N-terminal domain conjugated via additional C-terminal Cys to Diphtheria toxoid.
- Cat. no.:10782-558
- Supplier No.:R-1682-500
Specifications
About this item
Autosomal recessive mutations in DJ-1 cause early-onset familial Parkinson's disease. DJ-1 is considered a redox-sensitive cytoplasmic protein found in brain as well as other cell types.
Confirmed by WB using soluble mouse and human brain extracts, reactivity for major product diminished by peptide absorption.
Application Information:
WB and IHC. Suggested dilution of 1:5,000 is recommended for WB. DJ-1 is a soluble protein of 189 amino acids and detected with 23 kDa mobility by western blotting. The suggested dilution for IHC is 1:100. Detected astrocyte cytoplasmic labelling in human brain formaldehyde-treated tissue. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: DJ-1/PARK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal domain
Host: Rabbit
Isotype: IgG
Reactivity: