Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant SDF-1 A/CXCL12 (from E. coli)
 
undefined
Human Recombinant SDF-1 A/CXCL12 (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C prior to opening
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    bioactive, but no test is suitable to set an Acceptance Criteria (primary cell assay, in vivo assay or cell line is not commercially available)
  • Gene ID:
    P48061
  • Reconstitution instructions:
    Reconstitute with Sterile water at 0.1 mg/mL
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    CXCL12|PBSF
  • UniProtKB:
    P48061 (2)
  • Protein/peptide name:
    SDF-1alphaCXCL12
  • Purity:
    >95%
  • Molecular weight:
    8 kDa
  • Sequence:
    KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    Ambient

 

Frequently Bought Together