Human Recombinant GDF-15-D MUTANT (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Interleukin 8 (IL-8 or CXCL8) is a member of the CXC cytokine family and is produced by macrophages, epithelial, smooth muscle, and endothelial cells. IL-8 binds the G protein-coupled serpentine receptors CXCR1 and CXCR2. IL-8 recruits innate immune cells, induces phagocytosis, and stimulates angiogenesis.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: No test available, ED50 ≤Currently undetermined, bioactivity ≥Currently undetermined
- Gene ID:Q99988
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:MIC-1|Prostate differentiation factor|Placental TGF-β
- UniProtKB:Q99988
- Protein/peptide name:GDF-15-D MUTANT
- Purity:≥95%
- Molecular weight:12.4/24.8 kDa
- Sequence:MARNGDDCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLI QKTDTGVSLQ TYDDLLAKDC HCI
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together