Order Entry
Canada
ContactUsLinkComponent
Human Recombinant PDGF-AB (from E. coli)
Human Recombinant PDGF-AB (from E. coli)
Certificates
undefined
Human Recombinant PDGF-AB (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: NR6R-3T3 cell proliferation, ED50 ≤20 ng/ml, bioactivity ≥5.0×10⁴ units/mg
  • Gene ID:
    P04085/P01127
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    ODGF|GDGF
  • UniProtKB:
    P04085/P01127
  • Protein/peptide name:
    PDGF-AB
  • Purity:
    ≥95%
  • Molecular weight:
    Alpha: 14.4 kDa, beta: 12.4 kDa, total 26.8
  • Sequence:
    Alpha chain: MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT Beta chain: MSLGSLTIAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRPTQVQLRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETVAAARPVT
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together