Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IGF-I (from E. coli)
Mouse Recombinant IGF-I (from E. coli)
Certificates
undefined
Mouse Recombinant IGF-I (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: FDC-P1 cell proliferation, ED50 ≤20 ng/ml, bioactivity ≥5.0×10⁴ units/mg
  • Gene ID:
    Q8CAR0
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    mechano growth factor|IGF-IA|Somatomedin C
  • UniProtKB:
    Q8CAR0
  • Protein/peptide name:
    IGF-I
  • Purity:
    ≥95%
  • Molecular weight:
    7.7 kDa
  • Sequence:
    GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKAA
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together