Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant Rank Ligand (from E. coli)
Mouse Recombinant Rank Ligand (from E. coli)
Certificates
undefined
Mouse Recombinant Rank Ligand (from E. coli)
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity Assay: Ability to induce bioluminescence in engineered RANKL bioassay cells, ED₅₀ ≤50 ng/ml, Bioactivity ≥2.0×10⁴ units/mg
  • Gene ID:
    O35235
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    OPGL|TNFSF11|ODF|TRANCE
  • Protein/peptide name:
    Rank ligand
  • Purity:
    ≥95%
  • Molecular weight:
    19.5 kDa
  • Sequence:
    MPAMMEGSWLD VAQRGKPEAQ PFAHLTINAA SIPSGSHKVT LSSWYHDRGW AKISNMTLSN GKLRVNQDGF YYLYANICFR HHETSGSVPT DYLQLMVYVV KTSIKIPSSH NLMKGGSTKN WSGNSEFHFY SINVGGFFKL RAGEEISIQV SNPSLLDPDQ DATYFGAFKV QDID
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate and 20 mM Sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together