Human Recombinant FGF-21 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Fibroblast growth factor 21 (FGF-21) is an endocrine hormone that regulates energy homeostasis and exerts cardioprotective functions during heart injury. FGF-21 is expressed in the liver, pancreas, heart, and adipose tissues. FGF-21 signaling is activated through the FGFR1c receptor and β-Klotho co-receptor. FGF-21 is an important regulator of glucose uptake and reduces cell apoptosis under stress conditions.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NIH-3T3 proliferation with β-klotho and heparin, ED50 ≤500 ng/ml, Bioactivity ≥2.0 ×10³ unit/mg
- Gene ID:Q9NSA1
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:FGFL
- Protein/peptide name:FGF-21
- Purity:≥95%
- Molecular weight:19.5 kDa
- Sequence:MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together