Order Entry
Canada
ContactUsLinkComponent
Human Recombinant I-TAC (from E. coli)
Human Recombinant I-TAC (from E. coli)
Certificates
undefined
Human Recombinant I-TAC (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: Currently undetermined, ED50 ≤Currently undetermined, bioactivity ≥NA
  • Gene ID:
    O14625
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    B-R1|CXCL11
  • UniProtKB:
    O14625
  • Protein/peptide name:
    I-TAC
  • Purity:
    ≥95%
  • Molecular weight:
    8.3 kDa
  • Sequence:
    FPMFKRGRCL CIGPGVKAVK VADIEKASIM YPSNNCDKIE VIITLKENKG QRCLNPKSKQ ARLIIKKVER KNF
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Related Information

Frequently Bought Together