Anti-Canine distemper virus Mouse Monoclonal Antibody [clone: 8-1]
10673-312
: Abnova
- Antibody type:Primary
- Antigen name:Canine Distemper Virus
- Antigen symbol:CDV
- Clonality:Monoclonal
- Clone:8-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Virus
- Size:200 µg
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:CDV
- Storage buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage temperature:Store at 4°C.
- Immunogen:Envelope protein from canine distemper virus.
- Cat. no.:10673-312
- Supplier No.:MAB0393
Mouse monoclonal antibody raised against envelope protein from canine distemper virus.
Type: Primary
Antigen: Canine distemper virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 8-1
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Viruses