Anti-PDK4 Mouse Monoclonal Antibody [clone: 1A10]
Catalog # 10596-626
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase Kinase 4
- Antigen symbol:PDK4
- Clonality:Monoclonal
- Clone:1A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Size:100 µg
- Cross adsorption:No
- Gene ID:5166
- Antigen synonyms:FLJ40832|Pyruvate dehydrogenase kinase isoform 4|Pyruvate dehydrogenase kinase isozyme 4 mitochondrial.|Pyruvate dehydrogenase [lipoamide] kinase isozyme 4 mitochondrial|Pyruvate dehydrogenase kinase 4
- Storage temperature:Cool
- Shipping temperature:Ice
- Cat. no.:10596-626
- Supplier No.:H00005166-M04
Specifications
About this item
Type: Primary
Antigen: PDK4
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human