Diaphragm with 3 Double Slits of Different Widths
Catalog # 470163-768
Supplier: American 3B Scientific
Specifications
- Description:Diaphragm with 3 Double Slits of Different Widths
- Cat. no.:470163-768
- Supplier No.:U14100
Specifications
About this item
Photographic diffraction objects in a slide frame.
- Dimensions: 50x50 mm²
- Number of slits: 3 sets of 2
- Slit width [mm]: 0.10; 0.15; 0.20
- Slit spacing [mm]: 0.30