Eisco® Dynamics Cart System
470159-548
: Wards
- Description:Dynamics cart system
- Cat. no.:470159-548
- Supplier No.:DYSYS
An economical way to teach dynamics including position, velocity, acceleration, force, elastic and in elastic collisions, momentum, conservation of energy, and much more.
- Kit contains all the things you need to start to teach dynamics
If you are looking to get started teaching physics, or need to revamp your mechanics labs this is the perfect kit.
: The kits contains: 1× 1.2 m track with pulley, 1× standard trolley, 1× spring loaded trolley, 2x masses, 10×10 g masses, 1× ticker tape timer, 5× spare tapes, 1× pack of carbon discs, 1× reel of cotton, 1× spirit level 1× stopwatch.
Shipped in 2 boxes due to size.
Shipped in 2 boxes due to size.