ContactUsLinkComponent
Ward's® Chemistry Up In Vapor Demonstration
Ward's® Chemistry Up In Vapor Demonstration
Catalog # 470163-150
Supplier:  Ward's Science
undefined
Ward's® Chemistry Up In Vapor Demonstration
Catalog # 470163-150
Supplier:  Ward's Science
Supplier Number:  IS7033
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Ward's® Chemistry Up In Vapor Demonstration
  • Cat. no.:
    470163-150
  • Supplier No.:
    IS7033
  • No. of Uses:
    5
Specifications
About this item
"Genie in the tube."
  • Safely show a decomposition reaction in the classroom or lab
  • Demonstration can be performed 5 times with materials provided
Observe the dramatic catalytic decomposition of hydrogen peroxide to produce oxygen, water vapor, and an abundance of heat. This dramatic, large-scale demo illustrates concepts of catalysts, decomposition reactions, and exothermic reactions. Glass tube sold separately.
Ordering information: This kit includes hydrogen peroxide, sodium iodide, and an instruction manual with SDS. A glass tube is needed but not included.
Delivery: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.