Human Recombinant TRAIL (from E. coli)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a member of the tumor necrosis factor (TNF) family of cytokines. TRAIL is widely produced by a variety of cell types including tumor cells, smooth muscle of the lung and spleen, cerebellar glial cells, and thyroid follicular cells. TRAIL is a cytotoxic protein that induces apoptosis in tumor cells through the activation of the death receptors DR4 and DR5. TRAIL also binds the neutralizing decoy receptors, DcR1 and DcR2. Human TRAIL is active on mouse cells.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a member of the tumor necrosis factor (TNF) family of cytokines. TRAIL is widely produced by a variety of cell types including tumor cells, smooth muscle of the lung and spleen, cerebellar glial cells, and thyroid follicular cells. TRAIL is a cytotoxic protein that induces apoptosis in tumor cells through the activation of the death receptors DR4 and DR5. TRAIL also binds the neutralizing decoy receptors, DcR1 and DcR2. Human TRAIL is active on mouse cells.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
- Catalog No:
- 10787-192
- 10787-196
- 10822-412
- 10821-850
- 10787-194
- Protein/peptide name:
- TRAIL
- TRAIL
- TRAIL
- TRAIL
- TRAIL
- Protein synonyms:
- Apo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10
- Apo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10
- Apo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10
- Apo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10
- Apo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P50591
- P50591
- P50591
- P50591
- P50591
- Sequence:
- MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
- MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
- MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
- MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
- MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥90%
- ≥90%
- ≥90%
- ≥90%
- ≥90%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Molecular weight:
- 19.5 kDa
- 19.5 kDa
- 19.5 kDa
- 19.5 kDa
- 19.5 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Product Family Options
- Pack typeSizeOrigin
- : 100-115-10UG
Specifications
Cat. No.10787-192Protein/peptide nameTRAILProtein synonymsApo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP50591SequenceMRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVGProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity: No biological activity data is available at this timePurity≥90%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight19.5 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - : 100-115-50UG
Specifications
Cat. No.10787-196Protein/peptide nameTRAILProtein synonymsApo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP50591SequenceMRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVGProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity: No biological activity data is available at this timePurity≥90%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight19.5 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - : 100-115-100UG
Specifications
Cat. No.10822-412Protein/peptide nameTRAILProtein synonymsApo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP50591SequenceMRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVGProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity: No biological activity data is available at this timePurity≥90%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight19.5 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - : 100-115-500UG
Specifications
Cat. No.10821-850Protein/peptide nameTRAILProtein synonymsApo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP50591SequenceMRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVGProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity: No biological activity data is available at this timePurity≥90%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight19.5 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - : 100-115-1MG
Specifications
Cat. No.10787-194Protein/peptide nameTRAILProtein synonymsApo-2L|Apo-2 Ligand|CD253|TL2|TNFSF10Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP50591SequenceMRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVGProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity: No biological activity data is available at this timePurity≥90%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight19.5 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT



