ContactUsLinkComponent
Principles of Stoichiometry Lab Activity
Principles of Stoichiometry Lab Activity
Catalog # 470163-316
Supplier:  Ward's Science
undefined
Principles of Stoichiometry Lab Activity
Catalog # 470163-316
Supplier:  Ward's Science
Supplier Number:  IS8060
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Principles of Stoichiometry Lab Activity
  • No. of Lab Groups:
    5
  • Cat. no.:
    470163-316
  • Supplier No.:
    IS8060
Specifications
About this item
The Principles of Stoichiometry Lab Activity is designed for students to see what types of reactions occur when different solutions are mixed together.
  • Four Different Activities in One Kit
  • All Chemicals Required Included
  • Materials Minimize the Need for Teacher Preparation
  • Kit Includes Teacher's Guide
  • Students See What Happens When Various Solutions Are Mixed Together
Four different experiments focus on the concepts of decomposition reactions, single-replacement reactions, double-replacement reactions, and balanced reactions. The materials in this kit are packaged to minimize the need for teacher preparation. They provide five unique sets of materials for students working in groups of six.
The dropper-topped bottles allow students to perform the array of experiments without the risk of cross contamination of chemicals.
Ordering information: This kit includes cupric sulfate solution, cupric nitrate solution, nickel nitrate solution, lead nitrate solution, cobalt nitrate solution, nitric acid solution, iron(iii) nitrate solution, sodium carbonate solution, sodium sulfate solution, ammonium hydroxide solution, sodium chloride solution, delta solution, sodium iodide solution, sodium thiocyanate solution, sodium chromate solution, sodium thiocyanate solution, sodium hydroxide solution, deionized water, hydrochloric acid solution, nitric acid solution, sulfuric acid solution, calcium nitrate solution, sodium oxalate solution, aluminum metal strips, zinc metal strips, copper metal strips, magnesium metal strips, a preprinted acetate grid sheet, duel well combo plates, wooden dowels, forceps, carbon electrodes, cotton string, a teacher's guide, and a student copymaster.