ContactUsLinkComponent
Playing With Polymers Lab Activity
Playing With Polymers Lab Activity
Catalog # 470163-308
Supplier:  Ward's Science
undefined
Playing With Polymers Lab Activity
Catalog # 470163-308
Supplier:  Ward's Science
Supplier Number:  IS1002
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Playing with Polymers Lab Activity
  • No. of Lab Groups:
    6
  • Cat. no.:
    470163-308
  • Supplier No.:
    IS1002
Specifications
About this item
Learn How To Make Slime and Polymer Worms!
  • Two activities in one
  • All required chemicals are included
  • Perfect hands-on experience For young scientists
Teacher leads a class discussion about polymers and their importance in our everyday lives (specifically plastics). After the discussion students will be able to make two different polymers – worms and slime. 
Ordering information: Includes Sodium Alginate, Calcium Chloride, Polyvinyl Alcohol, Sodium Borate, Large Plastic Cups, Small Plastic Cups, and Teacher's Guide and Student Copymaster.