If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Separation of a Mixture: Physical Properties of Matter
- No. of Lab Groups:6
- Cat. no.:470163-288
- Supplier No.:IS1000
Specifications
About this item
Perfect Introduction to Physical Properties.
- Ideal for beginner scientists
- Separate a mixture based on only physical characteristics
- All chemicals required are included
After a class discussion of matter and the fact that matter may differ from other matter in a variety of ways, students will examine three different solids individually and observe some differences in physical properties. Sand, iron filings and salt will be observed and tested using water and magnets. Based on what they find individually they will come up with a method (as a class or as groups) to separate a mixture of all three.