Quantofix Nitrite/pH Test Strips, CTL Scientific Supply
10217-570
:
- Description:Nitrite/pH Test Strips
- Storage:RT
- Cat. no.:10217-570
- Range:NO2: 0-1-5-10-20-40-80 mg/L,
pH: 6.0-6.4-6.7-7.0-7.6-7.9-8.2-8.4-8.6 - Supplier No.:91338
Quantofix Nitrite/pH test strip for the semi-quantitative determination of Nitrite and pH in cooling lubricants
Quantofix Nitrite/pH has a gradation of scale: 0-1-5-10-20-40-80 mg/L NO2 - pH: 6.0-6.4-6.7-7.0-7.6-7.9-8.2-8.4-8.6.
The easy dip and read procedure provides a reliable result within 1 minute.
: Packaged as a box of 100.