Quantofix Lubricheck Test Strips, CTL Scientific Supply
Catalog # 10217-568
Supplier: Macherey-Nagel
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Lubricheck Test Strips
- Environmentally Preferable:
- Storage:RT
- Cat. no.:10217-568
- Scale:0-15-50-75-130-200 mmol/L KOH
- Supplier No.:91336
Specifications
About this item
Quantofix Lubricheck test strip for the semi-quantitative determination of the concentration of cooling lubricants
Quantofix Lubricheck has a gradation of scale: 0-15-50-75-130-200 mmol/L KOH.
The easy dip and read procedure provides a reliable result within 1 minute.
Ordering information: Packaged as a box of 100.