Quantofix Glucose Test Strips, CTL Scientific Supply
Catalog # 10217-518
Supplier: Macherey-Nagel
CAS Number:
Specifications
- Description:Glucose Test Strips
- Storage:RT
- Cat. no.:10217-518
- Range:0-5-100-250-500-1000-2000 mg/L
- Supplier No.:91348
Specifications
About this item
Quantofix Glucose Test strips for the semi-quantitative determination of glucose content.
Quantofix Glucose test strips has a gradation of scale: 0-5-100-250-500-1000-2000 mg/L glucose.
Ordering information: Packaged as a box of 100.