Quantofix Active Oxygen Test Strips, CTL Scientific Supply
Active Oxygen Test Strips, 0-4-8-15-25 mg/L
Supplier: Macherey-Nagel
: 91349
Quantofix Active Oxygen Test Strips, CTL Scientific Supply
Catalog # 10217-500
Supplier: Macherey-Nagel
CAS Number:
Jump to:
Product Details & Documents
Quantofix Active Oxygen Test trips for the semi-quantitative determination of active oxygen.
Quantofix Active Oxygen Test strips have a gradation of scale: 0-4-8-15-25 mg/L PMPS (potassium monopersulphate).
: Packaged as a box of 100.
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Active Oxygen Test Strips
- Range:0-4-8-15-25 mg/L
- Cat. No.:10217-500
- Storage:RT



