Order Entry
Canada
Orders LinkContactUsLinkComponent
Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply

Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply

Ascorbic Acid Test Strips, 0-50-100-200-300-500-1000-2000 mg/L
Catalog # 10217-492
Supplier:   Macherey-Nagel
Supplier #:  91314
Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply
Catalog # 10217-492
Supplier:  Macherey-Nagel
CAS Number:  
Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply
Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply

Ascorbic Acid Test Strips, 0-50-100-200-300-500-1000-2000 mg/L

CAS Number:  
Price
114,22 $
Log in for availability
Each
Product Details & Documents

Quantofix Ascorbic Acid Test strips for the semi-quantitative determination for vitamin C in solutions.

Quantofix Ascorbic Acid Test strips have a gradation of scale: 0-50-100-200-300-500-1000-2000 mg/L Vitamin C. Store test strips in a cool and dry place, protected from sunlight and moisture.

Ordering information: Packaged as a box of 100.

Documents

Specifications

Specifications

  • Description:
    Ascorbic Acid Test Strips
  • Range:
    0-50-100-200-300-500-1000-2000 mg/L
  • Cat. No.:
    10217-492
  • Storage:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.