If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Cryostat Lubrication Kit
- Cat. no.:10764-044
- Packaging:Metal Box
- Supplier No.:SPCLK04
Specifications
About this item
This product is a quality alternative to Leica and is made from the highest quality, low-temperature synthetic lubricants.
- Special low temperature food grade synthetic lubricants
- Tested to temperatures down to –70 °C
- Lubricants are safe and odorless
- Light oil perfect for for cross bearing assemblies and any shaft roller bearings
Low temperature synthetic grease is perfect for mating surfaces of the cutting block, cutting head moving parts or the internal sliding mechanisms within the microtome.