Order Entry
Canada
ContactUsLinkComponent
 
Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody [clone: 5A9F9] (HRP (Horseradish Peroxidase))
Catalog # 89495-100
Supplier:  Genscript
undefined
Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody [clone: 5A9F9] (HRP (Horseradish Peroxidase))
Catalog # 89495-100
Supplier:  Genscript
Supplier Number:  A01742-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    NWSHPQFEK Tag
  • Antigen symbol:
    NWSHPQFEK Tag
  • Clonality:
    Monoclonal
  • Clone:
    5A9F9
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Size:
    100 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS, pH 7.4, containing 10 mg/ml BSA and 0.01% thimerosal
  • Storage temperature:
    The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    NWSHPQFEK peptide conjugated to KLH
  • Purification:
    Protein A affinity column
  • Cat. no.:
    89495-100
  • Packaging:
    Vial
  • Supplier No.:
    A01742-100

Specifications

About this item

THE™ NWSHPQFEK Tag antibody [HRP], mAb, Mouse recognizes N-terminal and C-terminal Strep II tagged fusion proteins.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: