Order Entry
Canada
ContactUsLinkComponent
Anti-HSA Mouse Monoclonal Antibody [clone: 2C7B4]
Anti-HSA Mouse Monoclonal Antibody [clone: 2C7B4]
Catalog # 89494-406
Supplier:  Genscript
undefined
Anti-HSA Mouse Monoclonal Antibody [clone: 2C7B4]
Catalog # 89494-406
Supplier:  Genscript
Supplier Number:  A01731-40

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HSA
  • Antigen symbol:
    HSA
  • Clonality:
    Monoclonal
  • Clone:
    2C7B4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS, pH 7.4, containing 0.02% sodium azide
  • Storage temperature:
    The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    HSA protein
  • Purification:
    Protein A affinity column
  • Cat. no.:
    89494-406
  • Packaging:
    Vial
  • Supplier No.:
    A01731-40

Specifications

About this item

HSA Antibody (2C7B4), mAb, Mouse recognizes HSA proteins and it has no cross-reactivity with mouse serum albumin, rabbit serum albumin, BSA and OVA.

  • High specificit
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: HSA
Clonality: Monoclonal
Clone: 2C7B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: