Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: 5A8E5] (FITC (Fluorescein Isothiocyanate))
Catalog # 89495-076
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen name:DYKDDDDK Tag
- Antigen symbol:DYKDDDDK Tag
- Clonality:Monoclonal
- Clone:5A8E5
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Size:100 μg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 1% BSA and 0.02% sodium azide
- Storage temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:A synthetic peptide DYKDDDDK coupled to KLH
- Purification:Protein A affinity column
- Cat. no.:89495-076
- Packaging:Vial
- Supplier No.:A01632
Specifications
About this item
GenScript THE™ DYKDDDDK Tag antibody [FITC], mAb, Mouse recognizes C-terminal, N-terminal and internal DYKDDDDK tagged fusion proteins.
- Comprehensive pool
- Against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: