To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:c-Myc Tag
- Antigen symbol:c-Myc Tag
- Clonality:Monoclonal
- Clone:2G8D5
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Western blot:Yes
- Size:100 μg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:bHLHe39|Myc2|Nird|AU016757|Niard
- Storage buffer:PBS, pH 7.4, containing 1% BSA and 0.01% thimerosal
- Storage temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to residues 410-419 (EQKLISEEDL) coupled to KLH
- Purification:Protein A affinity column
- Cat. no.:89493-640
- Packaging:Vial
- Supplier No.:A00863-100
Specifications
About this item
GenScript THE™ c-Myc antibody [HRP], mAb, Mouse recognizes C-terminal, N-terminal and internal c-Myc-tagged fusion proteins expressed in prokaryotic or eukaryotic cells.It also recognizes the c-Myc gene in human cancers, such as Hela whole cell lysate.
- Comprehensive pool
- Against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: MYC
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: