Order Entry
Canada
ContactUsLinkComponent
Running Buffers and Reagents, Protein Electrophoresis, Bio-Rad
Running Buffers and Reagents, Protein Electrophoresis, Bio-Rad
Catalogue # 101643-376
Fournisseur:  Bio-Rad
Numéro CAS:  
Running Buffers and Reagents, Protein Electrophoresis, Bio-Rad
Catalogue # 101643-376
Fournisseur:  Bio-Rad
Supplier Number:  1610744
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Description:
    10x Tris/Tricine/SDS
  • Size:
    1 L
  • Cat. no.:
    101643-376
  • Supplier No.:
    1610744

Spécifications

À propos de cet article

Bio-Rad offers a range of running buffers and reagents for many types of gels including SDS PAGE, nondenaturing, isoelectric focusing (IEF), MOPS and MES for Bis-Tris gels, and Tris-Tricine

Zymogram renaturation and development buffers are also available.

Ordering information: For 10x Tris/glycine/SDS and 10x Tris/glycine running buffers, 5 L cubes offer convenience and economy. The cubes have easy-pouring spouts and are compact and stackable to save bench space.