- Description:Time and temperature sensitive label
- Dimensions:4×4"
- Cat. no.:10497-240
- Supplier No.:544657
In accordance with current IATA Chapter 17 revised regulations for temperature sensitive products, this package marking label is required by the TTTF (IATA - Time & Temperature Sensitive Task Force) effective July 1, 2012.
The marking label must be affixed to at least one side panel of each individual package. The lower half of the label may be used to indicate the transport temperature (allowable temperature range) in Celsius designation and in English. If you do not know the transport temperature range, the lower half of the label should be left blank.
In an effort to develop minimum standards for the air transport and handling of time and temperature sensitive products, it is the responsibility of the shipper to apply the time and temperature sensitive product label to the outer packaging visible to package handlers.